missing translation for 'onlineSavingsMsg'
Learn More
Learn More
USP30 Antibody - Azide and BSA Free, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP2-93487-0.1ml
This item is not returnable.
View return policy
Description
USP30 Polyclonal antibody specifically detects USP30 in Human, Mouse, Rat samples. It is validated for Western Blot, Immunocytochemistry/ Immunofluorescence
Specifications
| USP30 | |
| Polyclonal | |
| Western Blot 1:500-1:2000, Immunocytochemistry/ Immunofluorescence 1:50-1:100 | |
| Deubiquitinating enzyme 30, EC 3.1.2.15, EC 3.4.19.12, FLJ40511, MGC10702, ubiquitin carboxyl-terminal hydrolase 30, ubiquitin specific peptidase 30, ubiquitin specific protease 30, ubiquitin thioesterase 30, Ubiquitin thiolesterase 30, ubiquitin-specific protease 30, Ubiquitin-specific-processing protease 30, Ub-specific protease 30 | |
| Recombinant fusion protein containing a sequence corresponding to amino acids 57-390 of human USP30 (NP_116052.2). TERKKRRKGLVPGLVNLGNTCFMNSLLQGLSACPAFIRWLEEFTSQYSRDQKEPPSHQYLSLTLLHLLKALSCQEVTDDEVLDASCLLDVLRMYRWQISSFEEQDAHELFHVITSSLEDERDRQPRVTHLFDVHSLEQQSEITPKQITCRTRGSPHPTSNHWKSQHPFHGRLTSNMVCKHCEHQSPVRFDTFDSLSLSIPAATWGHPLTLDHCLHHFISSESVRDVVCDNCTKIEAKGTLNGEKVEHQRTTFVKQLKLGKLPQCLCIHLQRLSWSSHGTPLKRHEHVQFNEFLMMDIYKYHLLGHKPSQHNPKLNKNPGPTLELQDGPGAPTPV | |
| 0.1 mL | |
| Cancer, Cell Biology, Endocrinology, Signal Transduction | |
| 84749 | |
| Store at -20°C. Avoid freeze-thaw cycles. | |
| IgG |
| Western Blot, Immunofluorescence | |
| Unconjugated | |
| PBS (pH 7.3), 50% glycerol | |
| Rabbit | |
| Affinity purified | |
| RUO | |
| Primary | |
| Human, Mouse, Rat | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction