missing translation for 'onlineSavingsMsg'
Learn More
Learn More
USP29 Antibody - Azide and BSA Free, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP2-94672-0.1ml
This item is not returnable.
View return policy
Description
USP29 Polyclonal antibody specifically detects USP29 in Human samples. It is validated for Western Blot
Specifications
| USP29 | |
| Polyclonal | |
| Western Blot 1:500 - 1:1000 | |
| Deubiquitinating enzyme 29, EC 3.1.2.15, EC 3.4.19.12, HOM-TES-84/86, MGC163266, MGC163270, ubiquitin carboxyl-terminal hydrolase 29, ubiquitin specific peptidase 29, ubiquitin specific protease 29, ubiquitin thioesterase 29, Ubiquitin thiolesterase 29, ubiquitin-specific processing protease, Ubiquitin-specific-processing protease 29 | |
| A synthetic peptide corresponding to a sequence within amino acids 200-300 of human usp29 (NP_065954.1). LKYIQSNRKNPSSLEDLEKDRDLKLGPSFNTNCNGNPNLDETVLATQTLNAKNGLTSPLEPEHSQGDPRCNKAQVPLDSHSQQLQQGFPNLGNTCYMNAVL | |
| 0.1 mL | |
| Cell Biology | |
| 57663 | |
| Store at -20°C. Avoid freeze-thaw cycles. | |
| IgG |
| Western Blot | |
| Unconjugated | |
| PBS (pH 7.3), 50% glycerol | |
| Rabbit | |
| Affinity purified | |
| RUO | |
| Primary | |
| Human | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction