Learn More
Invitrogen™ USP28 Recombinant Rabbit Monoclonal Antibody (24GB2515)
Rabbit Recombinant Monoclonal Antibody
Brand:  Invitrogen™ MA557305
 View more versions of this product
Product Code. 30275984
  • £434.00 / 100µL
Description
USP28 Recombinant Monoclonal Antibody for Western Blot, ICC/IF, Flow
Modification of target proteins by ubiquitin participates in a wide array of biological functions. Proteins destined for degradation or processing via the 26 S proteasome are coupled to multiple copies of ubiquitin. However, attachment of ubiquitin or ubiquitin-related molecules may also result in changes in subcellular distribution or modification of protein activity. An additional level of ubiquitin regulation, deubiquitination, is catalyzed by proteases called deubiquitinating enzymes, which fall into four distinct families. Ubiquitin C-terminal hydrolases, ubiquitin-specific processing proteases (USPs),1 OTU-domain ubiquitin-aldehyde-binding proteins, and Jab1/Pad1/MPN-domain-containing metallo-enzymes. Among these four families, USPs represent the most widespread and represented deubiquitinating enzymes across evolution. USPs tend to release ubiquitin from a conjugated protein. They display similar catalytic domains containing conserved Cys and His boxes but divergent N-terminal and occasionally C-terminal extensions, which are thought to function in substrate recognition, subcellular localization, and protein-protein interactions.
TRUSTED_SUSTAINABILITY
Specifications
Specifications
USP28
Recombinant Monoclonal
Unconjugated
USP28
9830148O20Rik; AU022237; Deubiquitinating enzyme 28; Kiaa1515; mKIAA1515; ubiquitin carboxyl-terminal hydrolase 28; ubiquitin carboxyl-terminal hydrolase 28 variant 1; ubiquitin specific peptidase 28; ubiquitin specific protease 28; ubiquitin thioesterase 28; ubiquitin thiolesterase 28; ubiquitin-specific-processing protease 28; Usp28
Rabbit
Affinity chromatography
RUO
235323, 315639, 57646
-20°C
Liquid
Flow Cytometry, Western Blot, Immunocytochemistry
24GB2515
PBS with 50% glycerol and 0.02% sodium azide; pH 7.4
D3ZJ96, Q5I043, Q96RU2
USP28
A synthesized peptide derived from human USP28 (aa 1020-1077).
100 μL
Primary
Human, Mouse, Rat