Learn More
USP28 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand:  Proteintech 17707-1-AP-150UL
 View more versions of this product
Product Code. 16847644
  • £349.00 / 150µL
Description
WNK is a serine/threonine kinase that plays an important role in the regulation of electrolyte homeostasis, cell signaling, survival, and proliferation. Also, acts as an activator and inhibitor of sodium-coupled chloride cotransporters and potassium-coupled chloride cotransporters respectively. Activates SCNN1A, SCNN1B, SCNN1D, and SGK1. WNK localizes to a segment of kidney involved in salt, potassium, and pH homeostasis, the distal nephron. Mutations in the WNK gene result in Pseudohypaldosteronism and Neuropathy.
Specifications
Specifications
USP28
Polyclonal
Unconjugated
USP28
Deubiquitinating enzyme 28, KIAA1515, Ubiquitin thiolesterase 28, USP28
Rabbit
Antigen Affinity Chromatography
RUO
235323, 315639, 57646
-20°C
Liquid
Immunohistochemistry (Paraffin), Immunoprecipitation, Western Blot
0.38 mg/mL
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3
D3ZJ96, Q5I043, Q96RU2
Usp28
USP28 Fusion Protein Ag12003
150 μL
Primary
Human, Mouse, Rat