missing translation for 'onlineSavingsMsg'
Learn More
Learn More
USP25 Antibody - BSA Free, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP2-95136-0.1ml
This item is not returnable.
View return policy
Description
USP25 Polyclonal antibody specifically detects USP25 in Human, Mouse, Rat samples. It is validated for Western Blot
Specifications
| USP25 | |
| Polyclonal | |
| Western Blot 1:100 - 1:500 | |
| Deubiquitinating enzyme 25, EC 3.1.2.15, EC 3.4.19.12, ubiquitin carboxyl-terminal hydrolase 25, ubiquitin specific peptidase 25, ubiquitin specific protease USP2510USP21ubiquitin specific protease 25, ubiquitin thioesterase 25, Ubiquitin thiolesterase 25, ubiquitin-specific processing protease 25, Ubiquitin-specific-processing protease 25, USP on chromosome 21 | |
| Recombinant fusion protein containing a sequence corresponding to amino acids 1-230 of human USP25 (NP_037528.3). MTVEQNVLQQSAAQKHQQTFLNQLREITGINDTQILQQALKDSNGNLELAVAFLTAKNAKTPQQEETTYYQTALPGNDRYISVGSQADTNVIDLTGDDKDDLQRAIALSLAESNRAFRETGITDEEQAISRVLEASIAENKACLKRTPTEVWRDSRNPYDRKRQDKAPVGLKNVGNTCWFSAVIQSLFNLLEFRRLVLNYKPPSNAQDLPRNQKEHRNLPFMRELRYLFA | |
| 0.1 mL | |
| Cell Biology | |
| 29761 | |
| Store at -20°C. Avoid freeze-thaw cycles. | |
| IgG |
| Western Blot | |
| Unconjugated | |
| PBS (pH 7.3), 50% glycerol | |
| Rabbit | |
| Affinity purified | |
| RUO | |
| Primary | |
| Human, Mouse, Rat | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction