missing translation for 'onlineSavingsMsg'
Learn More
Learn More
USP25 Antibody, Alexa Fluor™ 594, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP2-82051AF594
This item is not returnable.
View return policy
Description
USP25 Polyclonal antibody specifically detects USP25 in Human, Mouse samples. It is validated for Western Blot, ELISA, Immunohistochemistry, Immunofluorescence, Immunohistochemistry (Paraffin)
Specifications
| USP25 | |
| Polyclonal | |
| 50mM Sodium Borate | |
| Rabbit | |
| Peptide affinity purified | |
| RUO | |
| Primary | |
| Human, Mouse | |
| Purified |
| Western Blot, ELISA, Immunohistochemistry, Immunofluorescence, Immunohistochemistry (Paraffin) | |
| Alexa Fluor 594 | |
| Deubiquitinating enzyme 25, EC 3.1.2.15, EC 3.4.19.12, ubiquitin carboxyl-terminal hydrolase 25, ubiquitin specific peptidase 25, ubiquitin specific protease USP2510USP21ubiquitin specific protease 25, ubiquitin thioesterase 25, Ubiquitin thiolesterase 25, ubiquitin-specific processing protease 25, Ubiquitin-specific-processing protease 25, USP on chromosome 21 | |
| USP25 antibody was raised against a 17 amino acid peptide near the center of human USP25. The immunogen is located within amino acids 470 - 520 of USP25. Amino Acid Squence: IPSQTLPSTTEQQGALS | |
| 0.1 mL | |
| Cell Biology | |
| 29761 | |
| Store at 4°C in the dark. | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction