missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ USP24 Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA568354
This item is not returnable.
View return policy
Description
Predicted to react with Mouse samples.
Modification of cellular proteins by ubiquitin is an essential regulatory mechanism controlled by the coordinated action of multiple ubiquitin-conjugating and deubiquitinating enzymes. USP24 belongs to a large family of cysteine proteases that function as deubiquitinating enzymes.
Specifications
| USP24 | |
| Polyclonal | |
| Unconjugated | |
| USP24 | |
| 2700066K03Rik; 2810030C21Rik; AI414051; B130021E18; C79851; deubiquitinating enzyme 24; KIAA1057; Ubiquitin carboxyl-terminal hydrolase 24; ubiquitin specific peptidase 24; ubiquitin specific protease 24; ubiquitin thioesterase 24; ubiquitin thiolesterase 24; ubiquitin-specific processing protease 24; ubiquitin-specific-processing protease 24; Usp24 | |
| Rabbit | |
| Affinity chromatography | |
| RUO | |
| 23358, 329908 | |
| -20°C | |
| Liquid |
| Western Blot | |
| 1 mg/mL | |
| PBS with 30% glycerol, 0.87% NaCl and 0.01% sodium azide; pH 7.3 | |
| B1AY13, Q9UPU5 | |
| USP24 | |
| Synthesized peptide derived from human USP24. | |
| 100 μL | |
| Primary | |
| Human, Mouse | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction