missing translation for 'onlineSavingsMsg'
Learn More
Learn More
USP24 Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP3-37932-20ul
This item is not returnable.
View return policy
Description
USP24 Polyclonal antibody specifically detects USP24 in Human samples. It is validated for ELISA,Western Blot
Specifications
| USP24 | |
| Polyclonal | |
| Western Blot 1:500 - 1:2000, ELISA | |
| Deubiquitinating enzyme 24, EC 3.4.19.12, FLJ31309, KIAA1057ubiquitin carboxyl-terminal hydrolase 24, ubiquitin specific peptidase 24, ubiquitin specific protease 24, ubiquitin thioesterase 24, Ubiquitin thiolesterase 24, ubiquitin-specific processing protease 24, Ubiquitin-specific-processing protease 24 | |
| Recombinant fusion protein containing a sequence corresponding to amino acids 2431-2620 of human USP24 (NP_056121.2).,, Sequence:, FTMLHFIKNQLETAPPHELKNTFQLLHEILVIEDPIQVERVKFVFETENGLLALMHHSNHVDSSRCYQCVKFLVTLAQKCPAAKEYFKENSHHWSWAVQWLQKKMSEHYWTPQSNVSNETSTGKTFQRTISAQDTLAYATALLNEKEQSGSSNGSESSPANENGDRHLQQGSESPMMIGELRSDLDDVDP | |
| 20 μL | |
| Cell Biology | |
| 23358 | |
| Store at -20°C. Avoid freeze-thaw cycles. | |
| IgG |
| ELISA, Western Blot | |
| Unconjugated | |
| PBS (pH 7.3), 50% glycerol | |
| Rabbit | |
| Affinity purified | |
| RUO | |
| Primary | |
| Human | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction