missing translation for 'onlineSavingsMsg'
Learn More
Learn More
USP22 Rabbit anti-Human, Mouse, Rat, Clone: 9C7I8, Novus Biologicals™
Rabbit Monoclonal Antibody
£158.00 - £386.00
Specifications
| Antigen | USP22 |
|---|---|
| Clone | 9C7I8 |
| Applications | Western Blot |
| Classification | Monoclonal |
| Conjugate | Unconjugated |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
18331847
|
Novus Biologicals
NBP3-16794-20UL |
20 μg |
£158.00
20µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
18332292
|
Novus Biologicals
NBP3-16794-100UL |
100 μg |
£386.00
100µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
USP22 Monoclonal antibody specifically detects USP22 in Human, Mouse, Rat samples. It is validated for Western BlotSpecifications
| USP22 | |
| Western Blot | |
| Unconjugated | |
| Rabbit | |
| Cancer, Cell Cycle and Replication, Signal Transduction, Stem Cell Markers | |
| PBS, 0.05% BSA, 50% glycerol, pH7.3 | |
| 23326 | |
| IgG | |
| Affinity purified |
| 9C7I8 | |
| Monoclonal | |
| Purified | |
| RUO | |
| Human, Mouse, Rat | |
| Deubiquitinating enzyme 22, EC 3.1.2.15, EC 3.4.19.12, KIAA1063ubiquitin carboxyl-terminal hydrolase 22, KIAA1064, ubiquitin specific peptidase 22, ubiquitin specific peptidase 3-like, ubiquitin specific protease 22, ubiquitin thioesterase 22, Ubiquitin thiolesterase 22, ubiquitin-specific processing protease 22, Ubiquitin-specific-processing protease 22, USP3L | |
| Recombinant fusion protein containing a sequence corresponding to amino acids 1-175 of human USP22 (Q9UPT9). | |
| Primary | |
| Store at -20°C. Avoid freeze-thaw cycles. |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title