missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ USP22 Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA512027
This item is not returnable.
View return policy
Description
This antibody is predicted to react with bovine, mouse and Xenopus and zebrafish based on sequence homology.
Histone deubiquitinating component of the transcription regulatory histone acetylation (HAT) complex SAGA. Catalyzes the deubiquitination of both histones H2A and H2B, thereby acting as a coactivator. Recruited to specific gene promoters by activators such as MYC, where it is required for transcription. Required for nuclear receptor-mediated transactivation and cell cycle progression.
Specifications
| USP22 | |
| Polyclonal | |
| PBS with 0.09% sodium azide; pH 7.4 | |
| Q9UPT9 | |
| USP22 | |
| KLH conjugated synthetic peptide between 351-380 amino acids from the C-terminal region of human USP22. | |
| 400 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
| Immunohistochemistry (Paraffin), Western Blot | |
| Unconjugated | |
| USP22 | |
| AI427806; Deubiquitinating enzyme 22; KIAA1063; Ubiquitin carboxyl-terminal hydrolase 22; ubiquitin specific peptidase 22; ubiquitin specific protease 22; ubiquitin thioesterase 22; ubiquitin thiolesterase 22; ubiquitin-specific processing protease 22; ubiquitin-specific-processing protease 22; Usp22; USP3L | |
| Rabbit | |
| Ammonium sulfate precipitation | |
| RUO | |
| 23326 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction