missing translation for 'onlineSavingsMsg'
Learn More
Learn More
USP21 Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP3-03947
This item is not returnable.
View return policy
Description
USP21 Polyclonal specifically detects USP21 in Human, Mouse samples. It is validated for Western Blot, Immunocytochemistry/Immunofluorescence.
Specifications
| USP21 | |
| Polyclonal | |
| PBS with 50% glycerol, pH7.3. | |
| Rabbit | |
| Affinity purified | |
| RUO | |
| Primary | |
| Human, Mouse | |
| Purified |
| Western Blot, Immunofluorescence | |
| Unconjugated | |
| Deubiquitinating enzyme 21, EC 3.1.2.15, EC 3.4.19.12, MGC3394, ubiquitin carboxyl-terminal hydrolase 21, ubiquitin specific peptidase 21, ubiquitin specific protease 21, ubiquitin thioesterase 21, Ubiquitin thiolesterase 21, ubiquitin-specific processing protease 21, ubiquitin-specific protease 16, Ubiquitin-specific-processing protease 21, USP16, USP23NEDD8-specific protease | |
| A synthetic peptide corresponding to a sequence within amino acids 400-500 of human USP21 (NP_001014443.1). | |
| 100 μL | |
| Cell Biology, Neuroscience, Ubiquitin Proteasome Pathway | |
| 27005 | |
| Store at -20°C. Avoid freeze-thaw cycles. | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction