Learn More
USP20 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand:  Proteintech 17491-1-AP-150UL
 View more versions of this product
Product Code. 16867384
  • £349.00 / 150µL
Description
SLAIN2 is closely related to SLAIN1, a novel stem cell protein that is expressed at the stem cell and epiblast stages. Unlike SLAIN1, SLAIN2 is widely expressed and is thought to play a role in regulating microtubule (MT) growth and organization. SLAIN2 binds to end-binding proteins (EBs), cytoplasmic linker proteins (CLIPs) such as CLIP170, CLIP-associated proteins, and interacts with the MT polymerase ch-TOG. Depletion of SLAIN2 using RNAi led to disorganization of the radial MT array, indicating its importance in the regulation of MT growth and organization.
Specifications
Specifications
USP20
Polyclonal
Unconjugated
USP20
Deubiquitinating enzyme 20, hVDU2, KIAA1003, LSFR3A, Ubiquitin thiolesterase 20, USP20, VDU2
Rabbit
Antigen Affinity Chromatography
RUO
10868, 311856, 74270
-20°C
Liquid
Immunohistochemistry (Paraffin), Immunoprecipitation, Western Blot
0.25 mg/mL
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3
Q8C6M1, Q9Y2K6
Usp20
USP20 Fusion Protein Ag11590
150 μL
Primary
Human, Mouse, Rat
Antibody
IgG
Suggestions de produits
Suggestions de produits
Vidéos
FDS
Documents
Documents
Certificats