missing translation for 'onlineSavingsMsg'
Learn More

USP20 Antibody, Novus Biologicals™

Product Code. 18323802 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
50 μg
Unit Size:
50µg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18323802 50 μg 50µg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 18323802 Supplier Novus Biologicals Supplier No. NBP314514

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Rabbit Polyclonal Antibody

USP20 Polyclonal antibody specifically detects USP20 in Human samples. It is validated for Immunohistochemistry, Immunohistochemistry (Paraffin)
TRUSTED_SUSTAINABILITY

Specifications

Antigen USP20
Applications Immunohistochemistry, Immunohistochemistry (Paraffin)
Classification Polyclonal
Concentration 1 mg/mL
Conjugate Unconjugated
Dilution Immunohistochemistry, Immunohistochemistry-Paraffin
Formulation PBS
Gene Alias Deubiquitinating enzyme 20, EC 3.4.19.12, KIAA1003hVDU2, LSFR3A, pVHL-interacting deubiquitinating enzyme 2, ubiquitin carboxyl-terminal hydrolase 20, ubiquitin specific peptidase 20, ubiquitin specific protease 20, ubiquitin thioesterase 20, Ubiquitin thiolesterase 20, Ubiquitin-specific-processing protease 20, VDU2, VHL-interacting deubiquitinating enzyme 2
Host Species Rabbit
Immunogen Synthetic 18 amino acid peptide from internal region of human USP20.
Purification Method Immunogen affinity purified
Quantity 50 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 10868
Target Species Human
Content And Storage Keep as concentrated solution. Aliquot and store at -20°C or below. Avoid multiple freeze-thaw cycles.
Product Type Antibody
Form Purified
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.