Learn More
USP2 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand:  Proteintech 10392-1-AP-20UL
 View more versions of this product
Product Code. 16846753
  • £135.00 / 20µL
Description
This antibody is specific to the isoform 1 (USP2a) of USP2.
Ubiquitin is a highly conserved protein involved in the regulation of intracellular protein breakdown, cell cycle regulation, and stress response, is released from degraded proteins by disassembly of the polyubiquitin chains. The disassembly process is mediated by ubiquitin-specific proteases (USPs).
Specifications
Specifications
USP2
Polyclonal
Unconjugated
USP2
Deubiquitinating enzyme 2, ubiquitin specific peptidase 2, Ubiquitin thiolesterase 2, UBP41, USP2, USP9
Rabbit
Antigen Affinity Chromatography
RUO
115771, 53376, 9099
-20°C
Liquid
Immunofluorescence, Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry
0.27 mg/mL
PBS with 50% glycerol and 0.1% sodium azide; pH 7.3
O75604, O88623, Q5U349
Usp2
USP2 Fusion Protein Ag0591
20 μL
Primary
Mouse, Rat, Human