missing translation for 'onlineSavingsMsg'
Learn More
Learn More
USP2 Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody
£305.00 - £410.00
Specifications
| Antigen | USP2 |
|---|---|
| Dilution | Western Blot 0.04 - 0.4 ug/ml, Immunohistochemistry 1:50 - 1:200, Immunocytochemistry/Immunofluorescence 0.25-2 ug/ml, Immunohistochemistry-Paraffin 1:20-1:50 |
| Classification | Polyclonal |
| Conjugate | Unconjugated |
| Host Species | Rabbit |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
18462821
|
Novus Biologicals
NBP1-86882-25ul |
25 μL |
£305.00
25µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
18221286
|
Novus Biologicals
NBP1-86882 |
0.1 mL |
£410.00
0.10mL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
USP2 Polyclonal specifically detects USP2 in Human samples. It is validated for Western Blot, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin.Specifications
| USP2 | |
| Polyclonal | |
| Rabbit | |
| Human, Mouse | |
| Deubiquitinating enzyme 2, EC 3.1.2.15, EC 3.4.19.12,41 kDa ubiquitin-specific protease, ubiquitin specific peptidase 2, ubiquitin specific protease 12, ubiquitin specific protease 2, ubiquitin specific protease 9, ubiquitin thioesterase 2, Ubiquitin thiolesterase 2, Ubiquitin-specific-processing protease 2, UBP41ubiquitin carboxyl-terminal hydrolase 2, USP9 | |
| USP2 | |
| IgG | |
| Affinity Purified | |
| Specificity of antibody verified on a Protein Array containing target protein plus 383 other non-specific proteins. |
| Western Blot 0.04 - 0.4 ug/ml, Immunohistochemistry 1:50 - 1:200, Immunocytochemistry/Immunofluorescence 0.25-2 ug/ml, Immunohistochemistry-Paraffin 1:20-1:50 | |
| Unconjugated | |
| RUO | |
| PBS (pH 7.2) and 40% Glycerol with 0.02% Sodium Azide | |
| 9099 | |
| This antibody was developed against Recombinant Protein corresponding to amino acids:SGFPYGVTNNCLSYLPINAYDQGVTLTQKLDSQSDLARDFSSLRTSDSYRIDPRNLGRSPMLARTRKELCTLQGLYQTASCPEYLVDYLENYGRKGSASQVPSQAPPSRVPEIISPTYR | |
| Primary | |
| Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. |
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title