missing translation for 'onlineSavingsMsg'
Learn More
Learn More
USP19 Antibody, HRP, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NB100-79828H
This item is not returnable.
View return policy
Description
USP19 Polyclonal antibody specifically detects USP19 in Human samples. It is validated for Western Blot
Specifications
| USP19 | |
| Polyclonal | |
| Western Blot | |
| Deubiquitinating enzyme 19, EC 3.1.2.15, EC 3.4.19.12, KIAA0891, ubiquitin carboxyl-terminal hydrolase 19, ubiquitin specific peptidase 19, ubiquitin thioesterase 19, Ubiquitin thiolesterase 19, Ubiquitin-specific-processing protease 19, Zinc finger MYND domain-containing protein 9, ZMYND9ubiquitin specific protease 19 | |
| The immunogen recognized by this antibody maps to a region between residue 1 and 50 of human ubiquitin specific peptidase 19 (zinc finger MYND domain-containing protein 9) using the numbering given in entry NP_006668.1 (GeneID 10869). | |
| 0.1 mL | |
| Neuroscience | |
| 10869 | |
| Store at 4C in the dark. | |
| IgG |
| Western Blot | |
| HRP | |
| PBS | |
| Rabbit | |
| Immunogen affinity purified | |
| RUO | |
| Primary | |
| Human | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction