missing translation for 'onlineSavingsMsg'
Learn More
Learn More
USP19 Antibody - Azide and BSA Free, Novus Biologicals™
Rabbit Polyclonal Antibody
£151.00 - £366.00
Specifications
| Antigen | USP19 |
|---|---|
| Dilution | Western Blot 1:500-1:2000, Immunocytochemistry/ Immunofluorescence 1:50-1:200 |
| Applications | Western Blot, Immunofluorescence |
| Classification | Polyclonal |
| Conjugate | Unconjugated |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
18638231
|
Novus Biologicals
NBP2-93529-0.02ml |
0.02 mL |
£151.00
0.02mL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
18686570
|
Novus Biologicals
NBP2-93529-0.1ml |
0.1 mL |
£366.00
0.10mL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
USP19 Polyclonal antibody specifically detects USP19 in Human, Mouse samples. It is validated for Western Blot, Immunocytochemistry/ ImmunofluorescenceSpecifications
| USP19 | |
| Western Blot, Immunofluorescence | |
| Unconjugated | |
| Rabbit | |
| Neuroscience | |
| PBS (pH 7.3), 50% glycerol | |
| 10869 | |
| IgG | |
| Affinity purified |
| Western Blot 1:500-1:2000, Immunocytochemistry/ Immunofluorescence 1:50-1:200 | |
| Polyclonal | |
| Purified | |
| RUO | |
| Human, Mouse | |
| Deubiquitinating enzyme 19, EC 3.1.2.15, EC 3.4.19.12, KIAA0891, ubiquitin carboxyl-terminal hydrolase 19, ubiquitin specific peptidase 19, ubiquitin thioesterase 19, Ubiquitin thiolesterase 19, Ubiquitin-specific-processing protease 19, Zinc finger MYND domain-containing protein 9, ZMYND9ubiquitin specific protease 19 | |
| Recombinant fusion protein containing a sequence corresponding to amino acids 1-200 of human USP19 (NP_001186089.1). MSGGASATGPRRGPPGLEDTTSKKKQKDRANQESKDGDPRKETGSRYVAQAGLEPLASGDPSASASHAAGITGSRHRTRLFFPSSSGSASTPQEEQTKEGACEDPHDLLATPTPELLLDWRQSAEEVIVKLRVGVGPLQLEDVDAAFTDTDCVVRFAGGQQWGGVFYAEIKSSCAKVQTRKGSLLHLTLPKKVPMLTWPS | |
| Primary | |
| Store at -20°C. Avoid freeze-thaw cycles. |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title