missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Specifications
Specifications
| Quantity | 0.1 mL |
| Content And Storage | Store at -80°C. Avoid freeze-thaw cycles. |
| Research Category | Neuroscience |
| Target | USP19 |
| Synonym | Deubiquitinating enzyme 19, EC 3.1.2.15, EC 3.4.19.12, KIAA0891, ubiquitin carboxyl-terminal hydrolase 19, ubiquitin specific peptidase 19, ubiquitin thioesterase 19, Ubiquitin thiolesterase 19, Ubiquitin-specific-processing protease 19, Zinc finger MYND domain-containing protein 9, ZMYND9ubiquitin specific protease 19 |
| Gene Symbol | USP19 |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?