missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ USP18 Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA5102960
This item is not returnable.
View return policy
Description
Antibody detects endogenous levels of total USP18.
USP18, a member of the deubiquitinating protease family of enzymes, removes ubiquitin adducts from a broad range of protein substrates.
Specifications
| USP18 | |
| Polyclonal | |
| Unconjugated | |
| USP18 | |
| 1110058H21Rik; 43 kDa ISG15-specific protease; AW047653; bubp43; hUBP43; ISG15-specific-processing protease; ISG43; putative ubiquitin-specific protease; ubiquitin specific peptidase 18; ubiquitin specific protease 15; ubiquitin specific protease 18; ubl carboxyl-terminal hydrolase 18; Ubl thioesterase 18; ubl thiolesterase 18; Ubp15; UBP43; USP18 | |
| Rabbit | |
| Affinity chromatography | |
| RUO | |
| 11274 | |
| -20°C | |
| Liquid |
| Western Blot | |
| 1 mg/mL | |
| PBS with 50% glycerol and 0.02% sodium azide; pH 7.4 | |
| Q9UMW8 | |
| USP18 | |
| A synthesized peptide derived from human USP18(Accession Q9UMW8), corresponding to amino acid residues L16-L66. | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction