missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ USP17L5 Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA5112883
This item is not returnable.
View return policy
Description
USP17L5 Polyclonal Antibody for Western Blot, IHC (P), ELISA
USP17L5 functions in cell apoptosis and cleaves ubiquitin fusion protein substrates.
Specifications
| USP17L5 | |
| Polyclonal | |
| Unconjugated | |
| USP17L5 | |
| Deubiquitinating enzyme 17-like protein 5; ubiquitin carboxyl-terminal hydrolase 17-like protein 5; ubiquitin specific peptidase 17-like 5; ubiquitin specific peptidase 17-like family member 5; ubiquitin thioesterase 17-like protein 5; ubiquitin thiolesterase 17-like protein 5; ubiquitin-specific-processing protease 17-like protein 5; USP17L5 | |
| Rabbit | |
| Protein G | |
| RUO | |
| 728386 | |
| -20°C or -80°C if preferred | |
| Liquid |
| ELISA, Immunohistochemistry (Paraffin), Western Blot | |
| 3.55 mg/mL | |
| PBS with 50% glycerol and 0.03% ProClin 300; pH 7.4 | |
| A8MUK1 | |
| USP17L5 | |
| Recombinant Human Ubiquitin carboxyl-terminal hydrolase 17-like protein 5 protein (19-235AA). | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction