missing translation for 'onlineSavingsMsg'
Learn More
Learn More
USP17L3 Polyclonal Antibody

Brand: Thermo Scientific PA5112880
This item is not returnable.
View return policy
Description
Specifications
| USP17L3 | |
| Polyclonal | |
| Unconjugated | |
| USP17L3 | |
| Deubiquitinating enzyme 17-like protein 3; ubiquitin carboxyl-terminal hydrolase 17-like protein 3; ubiquitin specific peptidase 17-like 3; ubiquitin specific peptidase 17-like family member 3; ubiquitin thioesterase 17-like protein 3; Ubiquitin-specific-processing protease 17-like protein 3; USP17B; USP17F; USP17L3 | |
| Rabbit | |
| Antigen Affinity Chromatography | |
| RUO | |
| 645836 | |
| -20°C or -80°C if preferred | |
| Liquid |
| ELISA, Western Blot | |
| 0.17 mg/mL | |
| PBS with 50% glycerol and 0.03% ProClin 300; pH 7.4 | |
| A6NCW0 | |
| USP17L3 | |
| Recombinant Human Ubiquitin carboxyl-terminal hydrolase 17-like protein 3 protein (19-251AA). | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction