missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ USP17L2 Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA544961
This item is not returnable.
View return policy
Description
Peptide sequence: FYIQKSEWER HSESVSRGRE PRALGAEDTD RRATQGELKR DHPCLQAPEL Sequence homology: Human: 100%.
DUB3 is a member of the ubiquitin processing protease (UBP) subfamily of deubiquitinating enzymes. See USP1 (MIM 603478) for background information.
Specifications
| USP17L2 | |
| Polyclonal | |
| Unconjugated | |
| USP17L2 | |
| deubiquitinating enzyme 17-like protein 2; deubiquitinating enzyme 3; deubiquitinating protein 3; DUB3; DUB-3; Ubiquitin carboxyl-terminal hydrolase 17; ubiquitin carboxyl-terminal hydrolase 17-like protein 2; ubiquitin specific peptidase 17-like 2; ubiquitin specific peptidase 17-like family member 2; ubiquitin thioesterase 17-like protein 2; ubiquitin thiolesterase 17-like protein 2; ubiquitin-specific-processing protease 17-like protein 2; USP17; USP17H; USP17I; USP17J; USP17K; USP17L; USP17L2; USP17M | |
| Rabbit | |
| Affinity Chromatography | |
| RUO | |
| 377630 | |
| -20°C, Avoid Freeze/Thaw Cycles | |
| Liquid |
| Western Blot | |
| 0.5 mg/mL | |
| PBS with 2% sucrose and 0.09% sodium azide | |
| Q6R6M4 | |
| USP17L2 | |
| Synthetic peptide directed towards the middle region of human USP17L2. | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction