missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ USP17L1 Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA5104196
This item is not returnable.
View return policy
Description
Antibody detects endogenous levels of total USP17L.
USP17L111 is a deubiquitinating enzyme that removes conjugated ubiquitin from specific proteins to regulate different cellular processes that may include cell proliferation, progression through the cell cycle, apoptosis, cell migration, and the cellular response to viral infection.
Specifications
| USP17L1 | |
| Polyclonal | |
| Unconjugated | |
| USP17L1 | |
| deubiquitinating enzyme 17-like protein 1; putative ubiquitin carboxyl-terminal hydrolase 17-like protein 1; ubiquitin carboxyl-terminal hydrolase 17-like protein 1; ubiquitin specific peptidase 17-like family member 1; ubiquitin thioesterase 17-like protein 1; Ubiquitin-specific-processing protease 17-like protein 1; USP17L; USP17L1; USP17L1P | |
| Rabbit | |
| Affinity chromatography | |
| RUO | |
| 401447 | |
| -20°C | |
| Liquid |
| Western Blot, Immunocytochemistry | |
| 1 mg/mL | |
| PBS with 50% glycerol and 0.02% sodium azide; pH 7.4 | |
| Q7RTZ2 | |
| USP17L1 | |
| A synthesized peptide derived from human USP17L1(Accession Q7RTZ2), corresponding to amino acid residues A367-A417. | |
| 100 μL | |
| Primary | |
| Human, Mouse, Rat | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction