missing translation for 'onlineSavingsMsg'
Learn More
Learn More
USP16 Antibody (OTI1B4), Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP2-46305
This item is not returnable.
View return policy
Description
Ensure accurate, reproducible results in Western Blot
USP16 Monoclonal specifically detects USP16 in Human, Mouse, Rat, Canine, Monkey samples. It is validated for Western Blot.
Specifications
| USP16 | |
| Monoclonal | |
| 1 mg/mL | |
| Western Blot 1:200 - 1:4000 | |
| Deubiquitinating enzyme 16, EC 3.1.2.15, EC 3.4.19.12, human ubiquitin processing protease, EC 3.1.2.1510ubiquitin specific protease 16, ubiquitin carboxyl-terminal hydrolase 16, ubiquitin specific peptidase 16, ubiquitin thioesterase 16, Ubiquitin thiolesterase 16, Ubiquitin-processing protease UBP-M, ubiquitin-specific processing protease 16, Ubiquitin-specific-processing protease 16, UBP-M | |
| Mouse | |
| Immunogen affinity purified | |
| RUO | |
| Primary | |
| Human, Mouse, Rat, Canine, Monkey | |
| Purified |
| Western Blot | |
| OTI1B4 | |
| Unconjugated | |
| PBS (pH 7.3), 1.0% BSA and 50% Glycerol with 0.02% Sodium Azide | |
| USP16 | |
| Human recombinant protein fragment corresponding to amino acids 137-466 of human USP16(NP_001001992) produced in E.coli. | |
| 0.1 mL | |
| Cell Cycle and Replication | |
| 10600 | |
| Store at -20C. Avoid freeze-thaw cycles. | |
| IgG2a |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction