missing translation for 'onlineSavingsMsg'
Learn More

USP16 Antibody (OTI1B4), Alexa Fluor™ 594, Novus Biologicals™

Product Code. 18423457 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
0.1 mL
Unit Size:
0.10mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18423457 0.1 mL 0.10mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 18423457 Supplier Novus Biologicals Supplier No. NBP274805AF594

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Mouse Monoclonal Antibody

USP16 Monoclonal antibody specifically detects USP16 in Human, Mouse, Rat, Canine, Monkey samples. It is validated for Western Blot
TRUSTED_SUSTAINABILITY

Specifications

Antigen USP16
Applications Western Blot
Classification Monoclonal
Clone OTI1B4
Conjugate Alexa Fluor 594
Formulation 50mM Sodium Borate
Gene Alias Deubiquitinating enzyme 16, EC 3.1.2.15, EC 3.4.19.12, human ubiquitin processing protease, EC 3.1.2.1510ubiquitin specific protease 16, ubiquitin carboxyl-terminal hydrolase 16, ubiquitin specific peptidase 16, ubiquitin thioesterase 16, Ubiquitin thiolesterase 16, Ubiquitin-processing protease UBP-M, ubiquitin-specific processing protease 16, Ubiquitin-specific-processing protease 16, UBP-M
Host Species Mouse
Immunogen Recombinant protein fragment corresponding to amino acids 137-466 of human USP16 (NP_001001992) produced in E.coli.
Purification Method Immunogen affinity purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Cell Cycle and Replication
Primary or Secondary Primary
Gene ID (Entrez) 10600
Target Species Human, Mouse, Rat, Canine, Monkey
Content And Storage Store at 4°C in the dark.
Form Purified
Isotype IgG2a
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.