missing translation for 'onlineSavingsMsg'
Learn More
Learn More
USP16 Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP3-38203-100ul
This item is not returnable.
View return policy
Description
USP16 Polyclonal antibody specifically detects USP16 in Human,Monkey,Mouse,Rat samples. It is validated for ELISA,Western Blot
Specifications
| USP16 | |
| Polyclonal | |
| Western Blot 1:500 - 1:1000, ELISA | |
| Deubiquitinating enzyme 16, EC 3.1.2.15, EC 3.4.19.12, human ubiquitin processing protease, EC 3.1.2.1510ubiquitin specific protease 16, ubiquitin carboxyl-terminal hydrolase 16, ubiquitin specific peptidase 16, ubiquitin thioesterase 16, Ubiquitin thiolesterase 16, Ubiquitin-processing protease UBP-M, ubiquitin-specific processing protease 16, Ubiquitin-specific-processing protease 16, UBP-M | |
| Recombinant fusion protein containing a sequence corresponding to amino acids 1-280 of human USP16 (NP_001001992.1).,, Sequence:, MGKKRTKGKTVPIDDSSETLEPVCRHIRKGLEQGNLKKALVNVEWNICQDCKTDNKVKDKAEEETEEKPSVWLCLKCGHQGCGRNSQEQHALKHYLTPRSEPHCLVLSLDNWSVWCYVCDNEVQYCSSNQLGQVVDYVRKQASITTPKPEKDNGNIELENKKLEKESKNEQEREKKENMAKENPPMNSPCQITVKGLSNLGNTCFFNAVMQNLSQTPVLRELLKEVKMSGTIVKIEPPDLALTEPLEINLEPPGPLTLAMSQFLNEMQETKKGVVTPKEL | |
| 100 μL | |
| Cell Cycle and Replication | |
| 10600 | |
| Store at -20°C. Avoid freeze-thaw cycles. | |
| IgG |
| ELISA, Western Blot | |
| Unconjugated | |
| PBS (pH 7.3), 50% glycerol | |
| Rabbit | |
| Affinity purified | |
| RUO | |
| Primary | |
| Human, Monkey, Mouse, Rat | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction