missing translation for 'onlineSavingsMsg'
Learn More
Learn More
USP15 Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody
£292.00 - £488.00
Specifications
| Antigen | USP15 |
|---|---|
| Applications | Immunocytochemistry, Immunofluorescence |
| Classification | Polyclonal |
| Conjugate | Unconjugated |
| Host Species | Rabbit |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
18288722
|
Novus Biologicals
NBP2-58023 |
100 μL |
£488.00
100µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
18618758
|
Novus Biologicals
NBP2-58023-25ul |
25 μL |
£292.00
25µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
USP15 Polyclonal specifically detects USP15 in Human samples. It is validated for Immunocytochemistry/Immunofluorescence.Specifications
| USP15 | |
| Polyclonal | |
| Rabbit | |
| DNA replication Transcription Translation and Splicing | |
| Deubiquitinating enzyme 15, EC 3.1.2.15, EC 3.4.19.12, KIAA0529ubiquitin carboxyl-terminal hydrolase 15, MGC131982, MGC149838, MGC74854, ubiquitin specific peptidase 15, ubiquitin specific protease 15, ubiquitin thioesterase 15, Ubiquitin thiolesterase 15, Ubiquitin-specific-processing protease 15, unph-2, UNPH4 | |
| USP15 | |
| IgG | |
| Affinity Purified |
| Immunocytochemistry, Immunofluorescence | |
| Unconjugated | |
| RUO | |
| Human | |
| 9958 | |
| This antibody was developed against a recombinant protein corresponding to the following amino acid sequence:DEPDDESSQDQELPSENENSQSEDSVGGDNDSENGLCTEDTCKGQLTGHKKRLFTFQFNNLGNTDINYIKDD | |
| Primary | |
| Specificity of antibody verified on a Protein Array containing target protein plus 383 other non-specific proteins. |
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title