missing translation for 'onlineSavingsMsg'
Learn More
Learn More
USP15 Antibody, HRP, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NB110-40690H
This item is not returnable.
View return policy
Description
USP15 Polyclonal antibody specifically detects USP15 in Human, Mouse samples. It is validated for Western Blot
Specifications
| USP15 | |
| Polyclonal | |
| Western Blot | |
| Deubiquitinating enzyme 15, EC 3.1.2.15, EC 3.4.19.12, KIAA0529ubiquitin carboxyl-terminal hydrolase 15, MGC131982, MGC149838, MGC74854, ubiquitin specific peptidase 15, ubiquitin specific protease 15, ubiquitin thioesterase 15, Ubiquitin thiolesterase 15, Ubiquitin-specific-processing protease 15, unph-2, UNPH4 | |
| The immunogen recognized by this antibody maps to a region between residue 960 and the C-terminus of human Ubiquitin Specific Peptidase 15 (AAD41086.1). | |
| 0.1 mL | |
| DNA replication Transcription Translation and Splicing | |
| 9958 | |
| Store at 4°C in the dark. | |
| IgG |
| Western Blot | |
| HRP | |
| PBS | |
| Rabbit | |
| Immunogen affinity purified | |
| RUO | |
| Primary | |
| Human, Mouse | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction