missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ USP14 Recombinant Rabbit Monoclonal Antibody (24GB2190)
Rabbit Recombinant Monoclonal Antibody
Brand: Invitrogen™ MA557817
This item is not returnable.
View return policy
Description
USP14 Recombinant Monoclonal Antibody for Western Blot, ICC/IF, Flow
This gene encodes a protein that is one of many involved in mechanisms to ensure proper chromosome segregation during cell division.
Specifications
| USP14 | |
| Recombinant Monoclonal | |
| Unconjugated | |
| Usp14 | |
| 2610005K12Rik; 2610037B11Rik; ataxia; AW107924; ax; C78769; deubiquitinating enzyme 14; dUB-type TGT for deubiquitinating enzyme; nmf375; TGT; tRNA-guanine transglycosylase, 60-kD subunit; ubiquitin carboxyl-terminal hydrolase 14; ubiquitin specific peptidase 14; ubiquitin specific peptidase 14 (tRNA-guanine transglycosylase); ubiquitin specific protease 14; ubiquitin specific protease 14 (tRNA-guanine transglycosylase); ubiquitin thioesterase 14; ubiquitin thiolesterase 14; ubiquitin-specific processing protease 14; ubiquitin-specific-processing protease 14; Usp14; wu:fk63d09; zgc:55949 | |
| Rabbit | |
| Affinity chromatography | |
| RUO | |
| 291796, 59025, 9097 | |
| -20°C | |
| Liquid |
| Flow Cytometry, Western Blot, Immunocytochemistry | |
| 24GB2190 | |
| PBS with 50% glycerol and 0.02% sodium azide; pH 7.4 | |
| P54578, Q9JMA1 | |
| Usp14 | |
| A synthesized peptide derived from human USP14 (aa 350-494). | |
| 20 μL | |
| Primary | |
| Human, Mouse, Rat | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction