missing translation for 'onlineSavingsMsg'
Learn More
Learn More
USP13 Antibody (OTI4G11), Alexa Fluor™ 700, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP2-74802AF700
This item is not returnable.
View return policy
Description
USP13 Monoclonal specifically detects USP13 in Human, Mouse, Rat, Canine, Monkey, Primate samples. It is validated for Western Blot, Immunohistochemistry.
Specifications
| USP13 | |
| Monoclonal | |
| Alexa Fluor 700 | |
| 50mM Sodium Borate with 0.05% Sodium Azide | |
| USP13 | |
| Full-length protein expressed in 293T cell transfected with human USP13 expression vector. | |
| 0.1 mL | |
| 8975.0 | |
| Store at 4°C in the dark. |
| Western Blot, Immunohistochemistry | |
| OTI4G11 | |
| Western Blot, Immunohistochemistry | |
| Deubiquitinating enzyme 13, EC 3.1.2.15, EC 3.4.19.12, Isopeptidase T-3, ISOT-3, ISOT3IsoT-3, ubiquitin carboxyl-terminal hydrolase 13, ubiquitin specific peptidase 13 (isopeptidase T-3), ubiquitin specific protease 13 (isopeptidase T-3), ubiquitin thioesterase 13, Ubiquitin thiolesterase 13, Ubiquitin-specific-processing protease 13 | |
| Mouse | |
| Affinity Purified | |
| Primary | |
| Human, Mouse, Rat, Canine, Monkey, Primate | |
| IgG1 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction