missing translation for 'onlineSavingsMsg'
Learn More
Learn More
USP13 Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP3-35117-20ul
This item is not returnable.
View return policy
Description
USP13 Polyclonal antibody specifically detects USP13 in Mouse,Rat samples. It is validated for ELISA,Western Blot
Specifications
| USP13 | |
| Polyclonal | |
| Western Blot 1:1000 - 1:2000, ELISA | |
| Deubiquitinating enzyme 13, EC 3.1.2.15, EC 3.4.19.12, Isopeptidase T-3, ISOT-3, ISOT3IsoT-3, ubiquitin carboxyl-terminal hydrolase 13, ubiquitin specific peptidase 13 (isopeptidase T-3), ubiquitin specific protease 13 (isopeptidase T-3), ubiquitin thioesterase 13, Ubiquitin thiolesterase 13, Ubiquitin-specific-processing protease 13 | |
| Recombinant fusion protein containing a sequence corresponding to amino acids 90-200 of human USP13 (NP_003931.2).,, Sequence:, MHLKRHVREKVRGASGGALPKRRNSKIFLDLDTDDDLNSDDYEYEDEAKLVIFPDHYEIALPNIEELPALVTIACDAVLSSKSPYRKQDPDTWENELPVSKYANNLTQLDN | |
| 20 μL | |
| Primary | |
| Mouse, Rat | |
| Purified |
| ELISA, Western Blot | |
| Unconjugated | |
| PBS (pH 7.3), 50% glycerol | |
| Rabbit | |
| Affinity purified | |
| RUO | |
| 8975 | |
| Store at -20°C. Avoid freeze-thaw cycles. | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction