missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ USP12 Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA5104193
This item is not returnable.
View return policy
Description
Antibody detects endogenous levels of total USP12.
Deubiquitinating enzyme. Has almost no deubiquitinating activity by itself and requires the interaction with WDR20 and WDR48 to have a high activity (PubMed:19075014, PubMed:27373336). Not involved in deubiquitination of monoubiquitinated FANCD2 (PubMed:19075014). In complex with WDR48, acts as a potential tumor suppressor by positively regulating PHLPP1 stability (PubMed:24145035). [UniProt].
Specifications
| USP12 | |
| Polyclonal | |
| Unconjugated | |
| USP12 | |
| Deubiquitinating enzyme 12; UBH1; Ubiquitin carboxyl-terminal hydrolase 12; ubiquitin hydrolyzing enzyme 1; ubiquitin specific peptidase 12; ubiquitin specific protease 12; ubiquitin specific protease 12 like 1; ubiquitin thioesterase 12; ubiquitin thiolesterase 12; ubiquitin-hydrolyzing enzyme 1; Ubiquitin-specific-processing protease 12; Usp12; USP12L1 | |
| Rabbit | |
| Affinity Chromatography | |
| RUO | |
| 219333, 22217, 360763 | |
| -20°C | |
| Liquid |
| Western Blot | |
| 1 mg/mL | |
| PBS with 50% glycerol and 0.02% sodium azide; pH 7.4 | |
| O75317, Q9D9M2 | |
| USP12 | |
| A synthesized peptide derived from human USP12(Accession O75317), corresponding to amino acid residues N134-T184. | |
| 100 μL | |
| Primary | |
| Human, Mouse, Rat | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction