missing translation for 'onlineSavingsMsg'
Learn More
Learn More
USP12 Antibody (OTI1E3), Alexa Fluor™ 647, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP2-74801AF647
This item is not returnable.
View return policy
Description
USP12 Monoclonal specifically detects USP12 in Human, Mouse, Rat samples. It is validated for Western Blot.
Specifications
| USP12 | |
| Monoclonal | |
| Alexa Fluor 647 | |
| 50mM Sodium Borate with 0.05% Sodium Azide | |
| USP12 | |
| Recombinant protein fragment corresponding to amino acids 61-251 of human USP12 (NP_872294) produced in E.coli. | |
| 0.1 mL | |
| 219333.0 | |
| Store at 4°C in the dark. |
| Western Blot | |
| OTI1E3 | |
| Western Blot | |
| Deubiquitinating enzyme 12, EC 3.1.2.15, EC 3.4.19.12, UBH1ubiquitin specific protease 12, ubiquitin carboxyl-terminal hydrolase 12, ubiquitin specific peptidase 12, ubiquitin specific protease 12 like 1, ubiquitin thioesterase 12, Ubiquitin thiolesterase 12, Ubiquitin-hydrolyzing enzyme 1, Ubiquitin-specific-processing protease 12, USP12L1 | |
| Mouse | |
| Affinity Purified | |
| Primary | |
| Human, Mouse, Rat | |
| IgG1 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction