Learn More
USP11 Rabbit anti-Human, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand:  Proteintech 22340-1-AP-20UL
 View more versions of this product
Product Code. 16871875
  • £135.00 / 20µL
Description
USP11 (ubiquitin specific peptidase 11), also known as UHX1, is a 920 amino acid deubiquitinating enzyme that participates in the Ub pathway. Localized to the nucleus, USP11 associates with both Ran BP-M (Ran binding protein M) and with the tumor suppressor BRCA2. Through these associations, USP11 functions to either inhibit ubiquitination of these proteins or to remove ubiquitin residues that have already been attached to these proteins. USP11 is implicated in several X-linked retinal diseases and, due to its ability to deubiquitinate BRCA2, may play a role in tumor suppression.
Specifications
Specifications
USP11
Polyclonal
Unconjugated
USP11
Deubiquitinating enzyme 11, Ubiquitin thiolesterase 11, UHX1, USP11
Rabbit
Antigen Affinity Chromatography
RUO
8237
-20°C
Liquid
Immunohistochemistry (Paraffin), Immunocytochemistry, Immunoprecipitation, Western Blot, Immunofluorescence
0.29 mg/mL
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3
P51784
USP11
USP11 Fusion Protein Ag17829
20 μL