missing translation for 'onlineSavingsMsg'
Learn More
Learn More
USP11 Rabbit anti-Human, Mouse, Rat, Clone: 8I1K10, Novus Biologicals™
Description
USP11 Monoclonal antibody specifically detects USP11 in Human, Mouse, Rat samples. It is validated for Western Blot
Specifications
Specifications
| Antigen | USP11 |
| Applications | Western Blot |
| Classification | Monoclonal |
| Clone | 8I1K10 |
| Conjugate | Unconjugated |
| Formulation | PBS, 0.05% BSA, 50% glycerol, pH7.3 |
| Gene Alias | Deubiquitinating enzyme 11, EC 3.1.2.15, EC 3.4.19.12, ubiquitin carboxyl-terminal hydrolase 11, ubiquitin specific peptidase 11, ubiquitin specific protease 11, ubiquitin thioesterase 11, Ubiquitin thiolesterase 11, ubiquitin-specific processing protease 11, Ubiquitin-specific-processing protease 11, UHX1ubiquitin carboxyl-terminal hydrolase, X-linked |
| Host Species | Rabbit |
| Immunogen | A synthetic peptide corresponding to a sequence within amino acids 800-900 of human USP11 (P51784). |
| Purification Method | Affinity purified |
| Show More |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?