missing translation for 'onlineSavingsMsg'
Learn More

USP11 Antibody, Novus Biologicals™

Rabbit Polyclonal Antibody

Brand:  Novus Biologicals NBP3-35921-100ul

 View more versions of this product

Product Code. 30226931

  • £417.00 / 100µL

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Description

Description

USP11 Polyclonal antibody specifically detects USP11 in Human,Mouse,Rat samples. It is validated for ELISA,Western Blot,Immunocytochemistry/Immunofluorescence
TRUSTED_SUSTAINABILITY
Specifications

Specifications

USP11
Polyclonal
Western Blot 1:500 - 1:1000, ELISA, Immunocytochemistry/Immunofluorescence 1:50 - 1:200
Deubiquitinating enzyme 11, EC 3.1.2.15, EC 3.4.19.12, ubiquitin carboxyl-terminal hydrolase 11, ubiquitin specific peptidase 11, ubiquitin specific protease 11, ubiquitin thioesterase 11, Ubiquitin thiolesterase 11, ubiquitin-specific processing protease 11, Ubiquitin-specific-processing protease 11, UHX1ubiquitin carboxyl-terminal hydrolase, X-linked
Recombinant fusion protein containing a sequence corresponding to amino acids 70-190 of human USP11 (NP_004642.2).,, Sequence:, PQHEELPGLDSQWRQIENGESGRERPLRAGESWFLVEKHWYKQWEAYVQGGDQDSSTFPGCINNATLFQDEINWRLKEGLVEGEDYVLLPAAAWHYLVSWYGLEHGQPPIERKVIELPNIQ
100 μL
Cell Biology, Direct Reversal of DNA Damage
8237
Store at -20°C. Avoid freeze-thaw cycles.
IgG
ELISA, Western Blot, Immunocytochemistry/Immunofluorescence
Unconjugated
PBS (pH 7.3), 50% glycerol
Rabbit
Affinity purified
RUO
Primary
Human, Mouse, Rat
Purified
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Promotions

Promotions

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.