missing translation for 'onlineSavingsMsg'
Learn More

USP10 Rabbit anti-Human, Mouse, Clone: 9L6F3, Novus Biologicals™

Product Code. 18386562
Change view
Click to view available options
Quantity:
100 μg
20 μg
Unit Size:
100µL
20µL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18386562 20 μg 20µL
18385785 100 μg 100µL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 18386562 Supplier Bio-Techne Supplier No. NBP31641620UL

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Rabbit Monoclonal Antibody

USP10 Monoclonal antibody specifically detects USP10 in Human, Mouse samples. It is validated for Western Blot, Immunofluorescence, Immunoprecipitation
TRUSTED_SUSTAINABILITY

Specifications

Antigen USP10
Applications Western Blot, Immunofluorescence, Immunoprecipitation
Classification Monoclonal
Clone 9L6F3
Conjugate Unconjugated
Formulation PBS, 0.05% BSA, 50% glycerol, pH7.3
Gene Alias Deubiquitinating enzyme 10, EC 3.4.19.12, KIAA0190MGC2621, ubiquitin carboxyl-terminal hydrolase 10, ubiquitin specific peptidase 10, ubiquitin specific protease 10, ubiquitin thioesterase 10, Ubiquitin thiolesterase 10, Ubiquitin-specific-processing protease 10, UBPO
Host Species Rabbit
Immunogen A synthetic peptide corresponding to a sequence within amino acids 1-100 of human USP10 (Q14694).
Purification Method Affinity purified
Quantity 20 μg
Regulatory Status RUO
Research Discipline Cell Biology
Primary or Secondary Primary
Gene ID (Entrez) 9100
Target Species Human, Mouse
Content And Storage Store at -20°C. Avoid freeze-thaw cycles.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.