missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ USP10 Polyclonal Antibody

Description
A suggested positive control is Jurkat cell lysate. PA5-20987 can be used with blocking peptide PEP-1101.
USP10, also known as ubiquitin specific peptidase 10, belongs to the ubiquitin-specific protease family of cysteine proteases. USP10 functions to catalyze the cleavage of ubiquitin from ubiquitin-conjugated protein substrates such as p53/TP53, SNX3 and CFTR. USP10 has been identified as a subunit of DNA-bound androgen receptor (AR) complexes and may play a role in the activity of the DNA-bound androgen receptor complex. USP10 also acts as an essential regulator of p53/TP53 stability and is thought to function as a tumor suppressor.
Specifications
Specifications
| Antigen | USP10 |
| Applications | Western Blot, Immunocytochemistry |
| Classification | Polyclonal |
| Concentration | 1 mg/mL |
| Conjugate | Unconjugated |
| Formulation | PBS with 0.02% sodium azide |
| Gene | Usp10 |
| Gene Accession No. | P52479, Q14694 |
| Gene Alias | 2610014N07Rik; Deubiquitinating enzyme 10; KIAA0190; MGC2621; mKIAA0190; Ode-1; ubiquintin c-terminal hydrolase related polypeptide; ubiquitin carboxyl-terminal hydrolase 10; ubiquitin specific peptidase 10; ubiquitin specific protease 10; ubiquitin thioesterase 10; ubiquitin thiolesterase 10; ubiquitin-specific-processing protease 10; UBPO; Uchrp; USP10 |
| Gene Symbols | Usp10 |
| Show More |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?