missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ USP10 Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA552334
This item is not returnable.
View return policy
Description
Immunogen sequence: KITPDGITKE ASYGSIDCQY PGSALALDGS SNVEAEVLEN DGVSGGLGQR ERKKKKKRPP GYYSYLKDGG DDSISTEALV NGHANSAVPN SVSAEDAEFM GDMPPPLTPR TCNSPQNSTD S Highest antigen sequence identity to the following orthologs: Mouse - 64%, Rat - 67%.
USP10, also known as ubiquitin specific peptidase 10, belongs to the ubiquitin-specific protease family of cysteine proteases. USP10 functions to catalyze the cleavage of ubiquitin from ubiquitin-conjugated protein substrates such as p53/TP53, SNX3 and CFTR. USP10 has been identified as a subunit of DNA-bound androgen receptor (AR) complexes and may play a role in the activity of the DNA-bound androgen receptor complex. USP10 also acts as an essential regulator of p53/TP53 stability and is thought to function as a tumor suppressor.
Specifications
| USP10 | |
| Polyclonal | |
| Unconjugated | |
| Usp10 | |
| 2610014N07Rik; Deubiquitinating enzyme 10; KIAA0190; MGC2621; mKIAA0190; Ode-1; ubiquintin c-terminal hydrolase related polypeptide; ubiquitin carboxyl-terminal hydrolase 10; ubiquitin specific peptidase 10; ubiquitin specific protease 10; ubiquitin thioesterase 10; ubiquitin thiolesterase 10; ubiquitin-specific-processing protease 10; UBPO; Uchrp; USP10 | |
| Rabbit | |
| Antigen affinity chromatography | |
| RUO | |
| 9100 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry | |
| 0.4 mg/mL | |
| PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
| Q14694 | |
| Usp10 | |
| Recombinant protein corresponding to Human USP10. Recombinant protein control fragment (Product #RP-89242). | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction