missing translation for 'onlineSavingsMsg'
Learn More

Usnic acid, Thermo Scientific Chemicals

Product Code. 15842534 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
1 g
5 g
25 g
Unit Size:
1g
25g
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
15842534 1 g 1g
15852534 5 g 5g
15862534 25 g 25g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 15842534 Supplier Thermo Scientific Acros Supplier No. 462850010
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 125-46-2
Molecular Formula C18H16O7
Molecular Weight (g/mol) 344.32
InChI Key CUCUKLJLRRAKFN-UHFFFAOYNA-N
IUPAC Name 4,10-diacetyl-11,13-dihydroxy-2,12-dimethyl-8-oxatricyclo[7.4.0.0²,⁷]trideca-1(13),6,9,11-tetraene-3,5-dione
SMILES CC(=O)C1C(=O)C=C2OC3=C(C(C)=O)C(O)=C(C)C(O)=C3C2(C)C1=O

Specifications

Melting Point 201.0°C to 203.0°C
Quantity 1 g
Formula Weight 344.32
Chemical Name or Material Usnic acid

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.