missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ USHBP1 Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA5100278
This item is not returnable.
View return policy
Description
Antibody detects endogenous levels of total MCC.
USHBP1, also named as Mutated in colon cancer protein 2 or AIE-75-binding protein, is a 703 amino acid protein, which belongs to the MCC family. Highest level of expression is in heart, and moderate to low expression in skeletal muscle, kidney, liver, small intestine, placenta and lung.
Specifications
| USHBP1 | |
| Polyclonal | |
| Unconjugated | |
| USHBP1 | |
| 2210404N08Rik; 9430064H24; AIE-75 binding protein; AIE-75-binding protein; AIEBP; MCC2; MCC-2; mutated in colon cancer protein 2; USH1 protein network component harmonin binding protein 1; USH1C-binding protein 1; USHBP1; Usher syndrome 1C binding protein 1; Usher syndrome type-1C protein-binding protein 1 | |
| Rabbit | |
| Affinity chromatography | |
| RUO | |
| 234395, 83878 | |
| -20°C | |
| Liquid |
| Western Blot | |
| 1 mg/mL | |
| PBS with 50% glycerol and 0.02% sodium azide; pH 7.4 | |
| Q8N6Y0, Q8R370 | |
| USHBP1 | |
| A synthesized peptide derived from human USHBP1(Accession Q8N6Y0), corresponding to amino acid residues L389-M439. | |
| 100 μL | |
| Primary | |
| Human, Mouse | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction