missing translation for 'onlineSavingsMsg'
Learn More

USH1C Antibody, FITC, Novus Biologicals™

Product Code. 18363863 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
0.1 mL
Unit Size:
0.10mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18363863 0.1 mL 0.10mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 18363863 Supplier Novus Biologicals Supplier No. NBP299408F

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Rabbit Polyclonal Antibody

USH1C Polyclonal specifically detects USH1C in Human samples. It is validated for ELISA, Immunohistochemistry, Immunohistochemistry-Paraffin.
TRUSTED_SUSTAINABILITY

Specifications

Antigen USH1C
Applications ELISA, Immunohistochemistry, Immunohistochemistry (Paraffin)
Classification Polyclonal
Conjugate FITC
Dilution ELISA, Immunohistochemistry, Immunohistochemistry-Paraffin
Formulation PBS
Gene Alias AIE75, AIE-75, Antigen NY-CO-38/NY-CO-37, Autoimmune enteropathy-related antigen AIE-75, deafness, autosomal recessive 18, DFNB18, harmonin, NY-CO-37, NY-CO-38, PDZ-45, PDZ73, PDZ-73, PDZ-73/NY-CO-38, Protein PDZ-73, Renal carcinoma antigen NY-REN-3, ush1cpst, Usher syndrome 1C (autosomal recessive, severe), Usher syndrome type-1C protein
Host Species Rabbit
Immunogen Produced in rabbits immunized with purified, recombinant USH1C (Q9Y6N9-1; Met 1-Phe 552).
Purification Method Affinity purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Signal Transduction, Vision
Primary or Secondary Primary
Gene ID (Entrez) 10083
Target Species Human
Content And Storage Store at 4°C in the dark.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.