missing translation for 'onlineSavingsMsg'
Learn More

USH1C Antibody (006), Alexa Fluor™ 700, Novus Biologicals™

Product Code. 18400358 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
0.1 mL
Unit Size:
0.10mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18400358 0.1 mL 0.10mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 18400358 Supplier Novus Biologicals Supplier No. NBP289584AF700

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Rabbit Monoclonal Antibody

USH1C Monoclonal antibody specifically detects USH1C in Human samples. It is validated for ELISA
TRUSTED_SUSTAINABILITY

Specifications

Antigen USH1C
Applications ELISA
Classification Monoclonal
Clone 006
Conjugate Alexa Fluor 700
Dilution ELISA
Formulation 50mM Sodium Borate
Gene Alias AIE75, AIE-75, Antigen NY-CO-38/NY-CO-37, Autoimmune enteropathy-related antigen AIE-75, deafness, autosomal recessive 18, DFNB18, harmonin, NY-CO-37, NY-CO-38, PDZ-45, PDZ73, PDZ-73, PDZ-73/NY-CO-38, Protein PDZ-73, Renal carcinoma antigen NY-REN-3, ush1cpst, Usher syndrome 1C (autosomal recessive, severe), Usher syndrome type-1C protein
Host Species Rabbit
Immunogen This antibody was obtained from a rabbit immunized with purified, recombinant Human USH1C (Q9Y6N9-1; Met1-Phe552).
Purification Method Protein A purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Signal Transduction, Vision
Primary or Secondary Primary
Gene ID (Entrez) 10083
Target Species Human
Content And Storage Store at 4°C in the dark.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.