missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ USF2 Polyclonal Antibody
Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA578217
This item is not returnable.
View return policy
Description
Positive Control: Jurkat, Raji, NCI-H929, NIH-3T3, Raw264.7, C2C12 Predicted Reactivity: Rat (97%), Xenopus laevis (81%), Bovine (97%) Store product as a concentrated solution. Centrifuge briefly prior to opening the vial.
USF2 consists of 43kDa (USF-1) and 44kDa (USF-2) polypeptides which independently exhibit site-specific DNA binding and are members of the c-Myc-related family of regulatory factors containing helix-loop-helix domains. USF also contains a leucine repeat that is required for efficient DNA binding.
Specifications
| USF2 | |
| Polyclonal | |
| Unconjugated | |
| USF2 | |
| bHLHb12; c-fos interacting; c-fos interacting protein; class B basic helix-loop-helix protein 12; FIP; FOS-interacting protein; Major late transcription factor 2; transcription factor USF2; upstream stimulatory factor 2; Upstream transcription factor 2; upstream transcription factor 2, c-fos interacting; USF2; Usf-2 | |
| Rabbit | |
| Antigen affinity chromatography | |
| RUO | |
| 22282, 7392 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| Western Blot, Immunocytochemistry | |
| 1 mg/mL | |
| PBS with 20% glycerol and 0.025% ProClin 300; pH 7 | |
| Q15853, Q64705 | |
| USF2 | |
| Recombinant protein encompassing a sequence within the center region of human USF2. The exact sequence is proprietary. | |
| 100 μL | |
| Primary | |
| Human, Mouse | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction