missing translation for 'onlineSavingsMsg'
Learn More
Learn More
USF2 Antibody, Allophycocyanin, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP1-92647APC
This item is not returnable.
View return policy
Description
USF2 Polyclonal antibody specifically detects USF2 in Human, Mouse samples. It is validated for Western Blot, Immunohistochemistry, Immunoprecipitation
Specifications
| USF2 | |
| Polyclonal | |
| Western Blot, Immunohistochemistry, Immunoprecipitation | |
| BHLHB12, bHLHb12upstream stimulatory factor 2, Class B basic helix-loop-helix protein 12, FIPc-fos interacting protein, FOS-interacting protein, Major late transcription factor 2, Upstream transcription factor 2, upstream transcription factor 2, c-fos interacting | |
| The immunogen this antibody was made to, maps to a region between residue 150 and 200 of human Upstream Transcription Factor 2 using the numbering given in entry NP_003358.1 (GeneID 7392). | |
| 0.1 mL | |
| Primary | |
| Human, Mouse | |
| Purified |
| Western Blot, Immunohistochemistry, Immunoprecipitation | |
| APC | |
| PBS | |
| Rabbit | |
| Immunogen affinity purified | |
| RUO | |
| 7392 | |
| Store at 4C in the dark. | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction