missing translation for 'onlineSavingsMsg'
Learn More

USF2 Antibody, Alexa Fluor™ 647, Novus Biologicals™

Product Code. 18400788 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
0.1 mL
Unit Size:
0.1mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18400788 0.1 mL 0.1mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 18400788 Supplier Novus Biologicals Supplier No. NBP192649AF647

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Rabbit Polyclonal Antibody

USF2 Polyclonal antibody specifically detects USF2 in Human, Mouse samples. It is validated for Western Blot, Immunohistochemistry, Immunohistochemistry (Paraffin)
TRUSTED_SUSTAINABILITY

Specifications

Antigen USF2
Applications Western Blot, Immunohistochemistry, Immunohistochemistry (Paraffin)
Classification Polyclonal
Conjugate Alexa Fluor 647
Dilution Western Blot, Immunohistochemistry, Immunohistochemistry-Paraffin
Formulation 50mM Sodium Borate
Gene Alias BHLHB12, bHLHb12upstream stimulatory factor 2, Class B basic helix-loop-helix protein 12, FIPc-fos interacting protein, FOS-interacting protein, Major late transcription factor 2, Upstream transcription factor 2, upstream transcription factor 2, c-fos interacting
Host Species Rabbit
Immunogen The immunogen this antibody was made to, maps to a region between residue 1 and 50 of human Upstream Transcription Factor 2 using the numbering given in entry NP_003358.1 (GeneID 7392).
Purification Method Immunogen affinity purified
Quantity 0.1 mL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 7392
Target Species Human, Mouse
Content And Storage Store at 4C in the dark.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.