missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ USF1 Recombinant Rabbit Monoclonal Antibody (6A0C9)
Rabbit Recombinant Monoclonal Antibody
Brand: Invitrogen™ MA555351
This item is not returnable.
View return policy
Description
Immunogen Sequence: TSDDAVDTEG TAAETHYTYF PSTAVGDGAG GTTSGSTAAV VTTQGSEALL GQATPPGTGQ FFVMMSPQEV LQGGSQRSIA PRTHPYSPKS EAPRTTRDEK R.
This gene encodes a member of the basic helix-loop-helix leucine zipper family, and can function as a cellular transcription factor. The encoded protein can activate transcription through pyrimidine-rich initiator (Inr) elements and E-box motifs. This gene has been linked to familial combined hyperlipidemia (FCHL). Two transcript variants encoding distinct isoforms have been identified for this gene.
Specifications
| USF1 | |
| Recombinant Monoclonal | |
| 0.2 mg/mL | |
| PBS with 0.05% BSA, 50% glycerol and 0.02% sodium azide; pH 7.3 | |
| P22415, Q61069 | |
| USF1 | |
| A synthetic peptide corresponding to a sequence within amino acids 100-200 of human USF1. | |
| 100 μL | |
| Primary | |
| Human, Mouse, Rat | |
| Antibody | |
| IgG |
| ELISA, Western Blot | |
| 6A0C9 | |
| Unconjugated | |
| USF1 | |
| bHLHb11; Class B basic helix-loop-helix protein 11; FCHL; FCHL1; HYPLIP1; Major late transcription factor 1; MLTF; MLTFI; UEF; Upstream stimulatory factor 1; upstream transcription factor 1; USF; USF1; usf1-bd | |
| Rabbit | |
| Protein A | |
| RUO | |
| 22278, 7391, 83586 | |
| -20°C, Avoid Freeze/Thaw Cycles | |
| Liquid |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction