missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ USF1 Recombinant Rabbit Monoclonal Antibody (24GB1890)
Rabbit Recombinant Monoclonal Antibody
Brand: Invitrogen™ MA557312
This item is not returnable.
View return policy
Description
USF1 Recombinant Monoclonal Antibody for Western Blot, ICC/IF, Flow
This gene encodes a member of the basic helix-loop-helix leucine zipper family, and can function as a cellular transcription factor. The encoded protein can activate transcription through pyrimidine-rich initiator (Inr) elements and E-box motifs. This gene has been linked to familial combined hyperlipidemia (FCHL). Two transcript variants encoding distinct isoforms have been identified for this gene.
Specifications
| USF1 | |
| Recombinant Monoclonal | |
| Unconjugated | |
| USF1 | |
| bHLHb11; Class B basic helix-loop-helix protein 11; FCHL; FCHL1; HYPLIP1; Major late transcription factor 1; MLTF; MLTFI; UEF; Upstream stimulatory factor 1; upstream transcription factor 1; USF; USF1; usf1-bd | |
| Rabbit | |
| Affinity chromatography | |
| RUO | |
| 7391, 83586 | |
| -20°C | |
| Liquid |
| Flow Cytometry, Western Blot, Immunocytochemistry | |
| 24GB1890 | |
| PBS with 50% glycerol and 0.02% sodium azide; pH 7.4 | |
| P22415 | |
| USF1 | |
| A synthesized peptide derived from human Upstream transcription factor 1 (aa 170-200). | |
| 20 μL | |
| Primary | |
| Human, Rat | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction