missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ USF1 Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA535031
This item is not returnable.
View return policy
Description
This antibody is predicted to react with bovine based on sequence homology.
This gene encodes a member of the basic helix-loop-helix leucine zipper family, and can function as a cellular transcription factor. The encoded protein can activate transcription through pyrimidine-rich initiator (Inr) elements and E-box motifs. This gene has been linked to familial combined hyperlipidemia (FCHL). Two transcript variants encoding distinct isoforms have been identified for this gene.
Specifications
| USF1 | |
| Polyclonal | |
| Unconjugated | |
| USF1 | |
| bHLHb11; Class B basic helix-loop-helix protein 11; FCHL; FCHL1; HYPLIP1; Major late transcription factor 1; MLTF; MLTFI; UEF; Upstream stimulatory factor 1; upstream transcription factor 1; USF; USF1; usf1-bd | |
| Rabbit | |
| Antigen affinity chromatography, Protein A | |
| RUO | |
| 22278, 7391 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| Flow Cytometry, Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry | |
| 0.5 mg/mL | |
| PBS with 0.09% sodium azide; pH 7.4 | |
| P22415, Q61069 | |
| USF1 | |
| KLH conjugated synthetic peptide between 174-201 amino acids from the central region of human USF1. | |
| 400 μL | |
| Primary | |
| Human, Mouse | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction