missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ USF1 Monoclonal Antibody (1264CT170.274.14)

Mouse Monoclonal Antibody
Brand: Invitrogen™ MA537637
This item is not returnable.
View return policy
Description
USF1 Monoclonal Antibody for Western Blot
This gene encodes a member of the basic helix-loop-helix leucine zipper family, and can function as a cellular transcription factor. The encoded protein can activate transcription through pyrimidine-rich initiator (Inr) elements and E-box motifs. This gene has been linked to familial combined hyperlipidemia (FCHL). Two transcript variants encoding distinct isoforms have been identified for this gene.
Specifications
| USF1 | |
| Monoclonal | |
| 0.5 mg/mL | |
| PBS with 0.09% sodium azide; pH 7.4 | |
| P22415 | |
| USF1 | |
| recombinant protein between 1-310 amino acids from the Central region of human USF1. | |
| 400 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG1 |
| Western Blot | |
| 1264CT170.274.14 | |
| Unconjugated | |
| USF1 | |
| bHLHb11; Class B basic helix-loop-helix protein 11; FCHL; FCHL1; HYPLIP1; Major late transcription factor 1; MLTF; MLTFI; UEF; Upstream stimulatory factor 1; upstream transcription factor 1; USF; USF1; usf1-bd | |
| Mouse | |
| Protein G | |
| RUO | |
| 7391 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction